Assay Design



Gene Transcripts

Gene Symbol
atp1a3a
Entrez Gene ID
64610
Gene Name
ATPase Na+/K+ transporting subunit alpha 3a
Gene Aliases
atp[a]3B, cb705, wu:fj56a06
Chromosome Location
Chr.19: 6328245 - 6385541 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131684.2 NP_571759.2 3 - 4 435 80
GenBank mRNA AF308599.1 - 3 - 4 361 80
BC064703.1 - 3 - 4 435 80

View Full Product Details