Assay Design



Gene Transcripts

Gene Symbol
atp1a1b
Entrez Gene ID
64616
Gene Name
ATPase Na+/K+ transporting subunit alpha 1b
Gene Aliases
atp1a1l5, atp[a]1B2, cb707, wu:fj34g01, zgc:92351
Chromosome Location
Chr.9: 259033 - 279558 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131690.1 NP_571765.1 16 - 17 2473 105
GenBank mRNA AY008375.1 - 16 - 17 2473 105
BC085663.1 - 16 - 17 2538 105

View Full Product Details