Assay Design



Gene Transcripts

Gene Symbol
vent
Entrez Gene ID
64810
Gene Name
ventral expressed homeobox
Gene Aliases
vega2, vent1, wu:fe36b11
Chromosome Location
Chr.13: 50609716 - 50614718 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131700.1 NP_571775.1 2 - 3 406 73
GenBank mRNA AF255044.1 - 2 - 3 406 73
AF277077.1 - 2 - 3 400 73

View Full Product Details