Assay Design



Gene Transcripts

Gene Symbol
eif2s1a
Entrez Gene ID
321807
Gene Name
eukaryotic translation initiation factor 2, subunit 1 alpha a
Gene Aliases
Eif2s1, eif2s1l, fb36a08, wu:fb36a08, zgc:56510
Chromosome Location
Chr.17: 34244345 - 34254233 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199569.1 NP_955863.1 2 - 3 409 64
GenBank mRNA BC049468.1 - 2 - 3 409 64
BC065313.1 - 2 - 3 358 64

View Full Product Details