Assay Design



Gene Transcripts

Gene Symbol
mylipa
Entrez Gene ID
335888
Gene Name
myosin regulatory light chain interacting protein a
Gene Aliases
Mir, mylip, wu:fj36b03, zgc:55987
Chromosome Location
Chr.19: 26340737 - 26378655 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199983.1 NP_956277.1 3 - 4 612 100
GenBank mRNA AY423020.1 - 3 - 4 612 100
AY434450.1 - 3 - 4 574 100
BC045844.1 - 3 - 4 600 100
BC065679.1 - 3 - 4 575 100

View Full Product Details