Assay Design



Gene Transcripts

Gene Symbol
sybl1
Entrez Gene ID
393236
Gene Name
synaptobrevin-like 1
Gene Aliases
zgc:56506
Chromosome Location
Chr.14: 127199 - 146570 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200266.1 NP_956560.1 3 - 4 407 101
XM_005157135.4 XP_005157192.1 3 - 4 425 101
GenBank mRNA BC049034.1 - 3 - 4 407 101

View Full Product Details