Assay Design



Gene Transcripts

Gene Symbol
mtmr8
Entrez Gene ID
393365
Gene Name
myotubularin related protein 8
Gene Aliases
si:dkey-286f3.1, zgc:56405
Chromosome Location
Chr.5: 20668 - 45134 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200394.1 NP_956688.1 9 - 10 1242 127
XM_021476036.1 XP_021331711.1 4 - 5 388 127
GenBank mRNA AY423041.1 - 9 - 10 1169 127
BC053411.1 - 9 - 10 1242 127

View Full Product Details