Assay Design



Gene Transcripts

Gene Symbol
mtmr8
Entrez Gene ID
393365
Gene Name
myotubularin related protein 8
Gene Aliases
si:dkey-286f3.1, zgc:56405
Chromosome Location
Chr.5: 20668 - 45134 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200394.1 NP_956688.1 13 - 14 1750 67
XM_021476036.1 XP_021331711.1 8 - 9 896 67
GenBank mRNA AY423041.1 - 13 - 14 1677 67
BC053411.1 - 13 - 14 1750 67

View Full Product Details