Assay Design
Gene Transcripts
- Gene Symbol
- gmppab
- Entrez Gene ID
-
393469
- Gene Name
- GDP-mannose pyrophosphorylase Ab
- Gene Aliases
-
gmppa, zgc:66135
- Chromosome Location
-
Chr.6: 13506858 - 13514230 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200497.1 |
NP_956791.1 |
8 - 9 |
1087 |
75 |
GenBank mRNA |
BC055506.1 |
- |
8 - 9 |
1087 |
75 |
View Full Product Details