Assay Design



Gene Transcripts

Gene Symbol
hnrnpl
Entrez Gene ID
394074
Gene Name
heterogeneous nuclear ribonucleoprotein L
Gene Aliases
hnrpl, zgc:55429
Chromosome Location
Chr.5: 141172 - 156962 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201099.1 NP_957393.1 7 - 8 761 62
GenBank mRNA BC045336.1 - 7 - 8 761 62
BC164693.1 - 7 - 8 773 62

View Full Product Details