Assay Design



Gene Transcripts

Gene Symbol
apmap
Entrez Gene ID
266637
Gene Name
adipocyte plasma membrane associated protein
Gene Aliases
bscv, cb351, wu:fb50a03, zgc:55833, zgc:85628
Chromosome Location
Chr.13: 6219624 - 6238930 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212608.1 NP_997773.1 7 - 8 949 101
GenBank mRNA BC044505.1 - 7 - 8 949 101
BC067549.1 - 7 - 8 970 101

View Full Product Details