Assay Design



Gene Transcripts

Gene Symbol
prpf6
Entrez Gene ID
323855
Gene Name
PRP6 pre-mRNA processing factor 6 homolog (S. cerevisiae)
Gene Aliases
c20orf14, fc12b02, wu:fa05f07, wu:fc12b02, zgc:65913
Chromosome Location
Chr.23: 7000173 - 7052386 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212655.2 NP_997820.2 10 - 11 1454 102
GenBank mRNA BC056710.1 - 10 - 11 102
BC066556.1 - 10 - 11 102

View Full Product Details