Assay Design



Gene Transcripts

Gene Symbol
arpc1b
Entrez Gene ID
335474
Gene Name
actin related protein 2/3 complex, subunit 1B
Gene Aliases
arpc1a, cb871, fj15h02, wu:fj08e12, wu:fj15h02, zgc:64056
Chromosome Location
Chr.3: 40809048 - 40820479 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213156.1 NP_998321.1 3 - 4 282 70
GenBank mRNA BC053229.1 - 3 - 4 282 70
BC164463.1 - 2 - 3 200 70

View Full Product Details