Assay Design



Gene Transcripts

Gene Symbol
naa10
Entrez Gene ID
406643
Gene Name
N-alpha-acetyltransferase 10, NatA catalytic subuni
Gene Aliases
ard1a, hm:zeh0223, wu:fc66b08, zgc:63981
Chromosome Location
Chr.23: 13832347 - 13840404 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213334.1 NP_998499.1 2 - 3 240 94
GenBank mRNA BC053180.1 - 2 - 3 240 94
BC165054.1 - 2 - 3 156 94

View Full Product Details