Assay Design



Gene Transcripts

Gene Symbol
b9d2
Entrez Gene ID
436667
Gene Name
B9 domain containing 2
Gene Aliases
zgc:92713
Chromosome Location
Chr.21: 21612137 - 21615651 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002394.1 NP_001002394.1 3 - 4 363 76
XM_005161162.4 XP_005161219.1 3 - 4 457 76
GenBank mRNA BC076193.1 - 3 - 4 363 76

View Full Product Details