Assay Design



Gene Transcripts

Gene Symbol
ttc33
Entrez Gene ID
436995
Gene Name
tetratricopeptide repeat domain 33
Gene Aliases
fb38g05, wu:fb38g05, zgc:86619
Chromosome Location
Chr.5: 251815 - 286298 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002722.1 NP_001002722.1 4 - 5 523 60
GenBank mRNA BC075750.1 - 4 - 5 523 60

View Full Product Details