Assay Design



Gene Transcripts

Gene Symbol
nitr1f
Entrez Gene ID
492323
Gene Name
novel immune-type receptor 1f
Gene Aliases
nitr1.33, nitr1.36, nitr1.49, nitr1f_1-2, si:busm1-219h16.3, si:dz219h16.3
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007292.1 NP_001007293.1 2 - 3 439 139

View Full Product Details