Assay Design



Gene Transcripts

Gene Symbol
pcyox1
Entrez Gene ID
550289
Gene Name
prenylcysteine oxidase 1
Gene Aliases
zgc:110246
Chromosome Location
Chr.10: 7653181 - 7671219 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017626.1 NP_001017626.1 2 - 3 270 69
GenBank mRNA BC092824.1 - 2 - 3 270 69
BC164221.1 - 1 - 2 145 69

View Full Product Details