Assay Design



Gene Transcripts

Gene Symbol
pin4
Entrez Gene ID
553574
Gene Name
protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin)
Gene Aliases
zgc:110008
Chromosome Location
Chr.14: 28432737 - 28437426 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020553.1 NP_001018389.1 2 - 3 357 69
GenBank mRNA BC095126.1 - 2 - 3 357 69
BC164671.1 - 2 - 3 149 69

View Full Product Details