Assay Design



Gene Transcripts

Gene Symbol
fbxo8
Entrez Gene ID
678549
Gene Name
F-box protein 8
Gene Aliases
zgc:136842
Chromosome Location
Chr.1: 38344657 - 38361460 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040307.1 NP_001035397.1 3 - 4 651 82
XM_021475840.1 XP_021331515.1 3 - 4 730 82
GenBank mRNA BC115288.1 - 3 - 4 687 82

View Full Product Details