Assay Design



Gene Transcripts

Gene Symbol
tnfrsf21
Entrez Gene ID
564874
Gene Name
tumor necrosis factor receptor superfamily, member 21
Gene Aliases
dr6, im:6795346, wu:fa55e01, wu:fb02e11, wu:fc29a09, wu:fi27h08
Chromosome Location
Chr.20: 35799912 - 35841581 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042688.2 NP_001036153.2 3 - 4 1528 64
GenBank mRNA BC141795.1 - 3 - 4 1528 64
DQ812118.1 - 3 - 4 1199 64

View Full Product Details