Assay Design



Gene Transcripts

Gene Symbol
scn12aa
Entrez Gene ID
795688
Gene Name
sodium channel, voltage gated, type XII, alpha a
Gene Aliases
-
Chromosome Location
Chr.2: 51887312 - 52045385 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044922.1 NP_001038387.1 22 - 23 4062 129
XM_009298901.3 XP_009297176.1 23 - 24 4582 129
GenBank mRNA DQ149507.1 - 22 - 23 4062 129

View Full Product Details