Assay Design



Gene Transcripts

Gene Symbol
zglp1
Entrez Gene ID
751739
Gene Name
zinc finger GATA like protein 1
Gene Aliases
zgc:153462
Chromosome Location
Chr.6: 153146 - 157389 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045449.1 NP_001038914.1 2 - 3 943 139
XM_021476933.1 XP_021332608.1 1 - 2 823 139
GenBank mRNA BC122298.1 - 2 - 3 943 139
BC155614.1 - - 1536 139

View Full Product Details