Assay Design



Gene Transcripts

Gene Symbol
srgap2
Entrez Gene ID
556198
Gene Name
SLIT-ROBO Rho GTPase activating protein 2
Gene Aliases
srgap2a, wu:fb47h12, zgc:136814
Chromosome Location
Chr.11: 243599 - 272700 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048235.1 NP_001041700.1 21 - 22 2945 74
XM_005172876.4 XP_005172933.1 21 - 22 2994 74
GenBank mRNA BC122886.1 - 21 - 22 2945 74

View Full Product Details