Assay Design



Gene Transcripts

Gene Symbol
ipmkb
Entrez Gene ID
790916
Gene Name
inositol polyphosphate multikinase b
Gene Aliases
zgc:158331
Chromosome Location
Chr.12: 7198761 - 7234915 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080064.1 NP_001073533.1 2 - 3 415 77
GenBank mRNA BC128841.1 - 2 - 3 415 77

View Full Product Details