Assay Design



Gene Transcripts

Gene Symbol
trim62.1
Entrez Gene ID
791192
Gene Name
tripartite motif containing 62, tandem duplicate 1
Gene Aliases
trim62, zgc:158653
Chromosome Location
Chr.11: 40331913 - 40419007 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080674.1 NP_001074143.1 3 - 4 778 93
XM_021479666.1 XP_021335341.1 3 - 4 810 93
GenBank mRNA BC129354.1 - 3 - 4 778 93

View Full Product Details