Assay Design



Gene Transcripts

Gene Symbol
nt5c1ba
Entrez Gene ID
798022
Gene Name
5'-nucleotidase, cytosolic IB a
Gene Aliases
zgc:163054
Chromosome Location
Chr.13: 32446169 - 32453932 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089444.1 NP_001082913.1 4 - 5 634 87
GenBank mRNA BC139706.1 - 4 - 5 634 87

View Full Product Details