Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098487.2 | NP_001091957.2 | 1 - 2 | 308 | 83 |
GenBank mRNA | AY216588.1 | - | 1 - 2 | 231 | 83 |
BC065465.1 | - | 1 - 2 | 279 | 83 | |
BC095658.1 | - | 1 - 2 | 273 | 83 | |
CD285269.1 | - | 1 - 2 | 308 | 83 |