Assay Design



Gene Transcripts

Gene Symbol
casp8l1
Entrez Gene ID
100049706
Gene Name
caspase 8, apoptosis-related cysteine peptidase, like 1
Gene Aliases
casp8b
Chromosome Location
Chr.6: 13095457 - 13102003 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098619.1 NP_001092089.1 3 - 4 348 82
XM_005165822.4 XP_005165879.2 4 - 5 1204 82
GenBank mRNA DQ812121.1 - 3 - 4 348 82
MG958001.1 - 3 - 4 348 82

View Full Product Details