Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130962.1 | NP_571037.1 | 2 - 3 | 417 | 81 |
XM_005165264.4 | XP_005165321.1 | 2 - 3 | 455 | 81 | |
XM_021475997.1 | XP_021331672.1 | 2 - 3 | 458 | 81 | |
GenBank mRNA | BC060892.1 | - | 2 - 3 | 381 | 81 |
D21135.1 | - | 2 - 3 | 417 | 81 |