Assay Design



Gene Transcripts

Gene Symbol
nipsnap2
Entrez Gene ID
30232
Gene Name
nipsnap homolog 2
Gene Aliases
gbas, wu:fc08b08, zgc:92497
Chromosome Location
Chr.15: 404875 - 414036 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131034.1 NP_571109.1 - 632 77
GenBank mRNA AJ249797.1 - - 622 77
BC081660.1 - - 632 77

View Full Product Details