Assay Design
Gene Transcripts
- Gene Symbol
- pla2g4aa
- Entrez Gene ID
-
30554
- Gene Name
- phospholipase A2, group IVAa (cytosolic, calcium-dependent)
- Gene Aliases
-
PLA2G4A, cpla2, pla2g4
- Chromosome Location
-
Chr.2: 20925899 - 20957362 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_131295.2 |
NP_571370.1 |
15 - 16 |
1844 |
75 |
GenBank mRNA |
BC162082.1 |
- |
14 - 15 |
1772 |
75 |
U10330.1 |
- |
15 - 16 |
1845 |
75 |
View Full Product Details