Assay Design



Gene Transcripts

Gene Symbol
fzd9b
Entrez Gene ID
58023
Gene Name
frizzled class receptor 9b
Gene Aliases
fz11, fzd9, fzx, hm:zehl0603, zehl0603, zg11
Chromosome Location
Chr.18: 163058 - 164803 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131511.1 NP_571586.1 1 - 1 1654 88
GenBank mRNA AF169639.1 - 1 - 1 1654 88
BC163365.1 - 1 - 1 1718 88

View Full Product Details