Assay Design



Gene Transcripts

Gene Symbol
tbx1
Entrez Gene ID
368206
Gene Name
T-box transcription factor 1
Gene Aliases
mp:zf637-3-000616, zgc:136724
Chromosome Location
Chr.5: 15201823 - 15215479 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183339.1 NP_899182.1 3 - 4 1032 102
XM_005168611.4 XP_005168668.1 4 - 5 1461 102
XM_009301230.3 XP_009299505.1 4 - 5 1408 102
GenBank mRNA AY151388.1 - 3 - 4 1032 102
AY197606.1 - 3 - 4 672 102
AY277631.1 - 3 - 4 802 102
AY294284.1 - 3 - 4 1032 102
BC115243.1 - 3 - 4 1026 102

View Full Product Details