Assay Design



Gene Transcripts

Gene Symbol
vax1
Entrez Gene ID
373870
Gene Name
ventral anterior homeobox 1
Gene Aliases
-
Chromosome Location
Chr.17: 110236 - 118951 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194410.2 NP_919391.2 2 - 3 640 57
GenBank mRNA AY185348.1 - 2 - 3 408 57
BC065951.1 - 2 - 3 640 57

View Full Product Details