Assay Design



Gene Transcripts

Gene Symbol
slc43a1a
Entrez Gene ID
334395
Gene Name
solute carrier family 43 member 1a
Gene Aliases
fi47a12, lat3a, slc43a1, wu:fi47a12, zgc:55850
Chromosome Location
Chr.14: 258615 - 280030 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001327986.1 NP_001314915.1 10 - 11 1270 67
GenBank mRNA BC044509.1 - 10 - 11 1235 67
BC164469.1 - 9 - 10 992 67

View Full Product Details