Assay Design
Gene Transcripts
- Gene Symbol
- cnppd1
- Entrez Gene ID
-
334868
- Gene Name
- cyclin Pas1/PHO80 domain containing 1
- Gene Aliases
-
c2orf24, wu:fa05h07, zgc:55857
- Chromosome Location
-
Chr.1: 6161989 - 6169817 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_199928.1 |
NP_956222.1 |
2 - 3 |
262 |
63 |
GenBank mRNA |
BC044514.1 |
- |
2 - 3 |
262 |
63 |
View Full Product Details