Assay Design
Gene Transcripts
- Gene Symbol
- pip4k2ca
- Entrez Gene ID
-
373124
- Gene Name
- phosphatidylinositol-5-phosphate 4-kinase, type II, gamma a
- Gene Aliases
-
Pip5k2c, cb770, pip4k2c, pip5k2, zgc:56046
- Chromosome Location
-
Chr.23: 36616854 - 36645156 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200101.2 |
NP_956395.2 |
6 - 7 |
697 |
145 |
GenBank mRNA |
BC071466.1 |
- |
6 - 7 |
697 |
145 |
View Full Product Details