Assay Design
Gene Transcripts
- Gene Symbol
- pole3
- Entrez Gene ID
-
393774
- Gene Name
- polymerase (DNA directed), epsilon 3 (p17 subunit)
- Gene Aliases
-
zgc:73351
- Chromosome Location
-
Chr.5: 389853 - 392328 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200801.1 |
NP_957095.1 |
1 - 2 |
355 |
69 |
GenBank mRNA |
BC059666.1 |
- |
1 - 2 |
355 |
69 |
View Full Product Details