Assay Design
Gene Transcripts
- Gene Symbol
- fbxo30a
- Entrez Gene ID
-
100001417
- Gene Name
- F-box protein 30a
- Gene Aliases
-
fbxo30, zgc:63656
- Chromosome Location
-
Chr.17: 7620240 - 7628458 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_201151.1 |
NP_957445.1 |
2 - 3 |
2118 |
116 |
GenBank mRNA |
BC055189.1 |
- |
2 - 3 |
2118 |
116 |
View Full Product Details