Assay Design



Gene Transcripts

Gene Symbol
aar2
Entrez Gene ID
431768
Gene Name
AAR2 splicing factor
Gene Aliases
im:7153138, zgc:92018
Chromosome Location
Chr.23: 4216193 - 4225606 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002221.1 NP_001002221.1 2 - 3 785 102
XM_005161924.4 XP_005161981.1 3 - 4 895 102
GenBank mRNA BC072724.1 - 2 - 3 785 102

View Full Product Details