Assay Design



Gene Transcripts

Gene Symbol
gcshb
Entrez Gene ID
436852
Gene Name
glycine cleavage system protein H (aminomethyl carrier), b
Gene Aliases
wu:fd56h06, zgc:110557, zgc:92732
Chromosome Location
Chr.7: 67451169 - 67463021 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002579.1 NP_001002579.1 1 - 2 218 74
GenBank mRNA AY847710.1 - 1 - 2 169 74
BC076212.1 - 1 - 2 218 74

View Full Product Details