Assay Design



Gene Transcripts

Gene Symbol
syf2
Entrez Gene ID
445043
Gene Name
SYF2 pre-mRNA-splicing factor
Gene Aliases
hm:zeh0031, zeh0031, zgc:113871, zgc:92835
Chromosome Location
Chr.13: 45431695 - 45443008 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003437.2 NP_001003437.1 3 - 4 259 69
GenBank mRNA BC076298.1 - 3 - 4 260 69

View Full Product Details