Assay Design



Gene Transcripts

Gene Symbol
ccdc80
Entrez Gene ID
368419
Gene Name
coiled-coil domain containing 80
Gene Aliases
CL2, SSG-1, si:bz20i5.2, ssg1, zgc:113236
Chromosome Location
Chr.9: 34089156 - 34113209 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007198.1 NP_001007199.1 5 - 6 2201 63
GenBank mRNA BC090295.1 - 2 - 3 376 63
BC171523.1 - 5 - 6 2267 63
BC171525.1 - 5 - 6 2267 63
EU269066.1 - 5 - 6 2390 63

View Full Product Details