Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014310.2 | NP_001014332.2 | 3 - 4 | 350 | 60 |
XM_021468818.1 | XP_021324493.1 | 3 - 4 | 353 | 60 | |
GenBank mRNA | BC129488.1 | - | 3 - 4 | 350 | 60 |
BC165857.1 | - | 2 - 3 | 291 | 60 | |
KF270628.1 | - | 2 - 3 | 257 | 60 |