Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014822.1 | NP_001014822.1 | 17 - 18 | 1960 | 145 |
XM_005166039.3 | XP_005166096.1 | 18 - 19 | 2505 | 145 | |
XM_005166040.3 | XP_005166097.1 | 17 - 18 | 2415 | 145 | |
GenBank mRNA | AY766118.1 | - | 17 - 18 | 1960 | 145 |
BC163764.1 | - | 17 - 18 | 2016 | 145 | |
BC163771.1 | - | 17 - 18 | 2016 | 145 | |
CU458859.1 | - | 17 - 18 | 1975 | 145 |