Assay Design



Gene Transcripts

Gene Symbol
lin7b
Entrez Gene ID
553768
Gene Name
lin-7 homolog B (C. elegans)
Gene Aliases
fj41b11, fj63g03, wu:fj41b11, wu:fj63g03, zgc:109997
Chromosome Location
Chr.3: 31933925 - 31944709 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020733.2 NP_001018569.2 3 - 4 394 79
GenBank mRNA AY584746.1 - 3 - 4 394 79
BC095116.1 - 3 - 4 426 79
BC165891.1 - 3 - 4 260 79

View Full Product Details