Assay Design



Gene Transcripts

Gene Symbol
nom1
Entrez Gene ID
557585
Gene Name
nucleolar protein with MIF4G domain 1
Gene Aliases
si:ch211-266o5.1
Chromosome Location
Chr.7: 120042 - 132961 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044832.3 NP_001038297.3 8 - 9 1123 109
NM_001328060.1 NP_001314989.1 8 - 9 2094 109
GenBank mRNA BC134855.1 - 8 - 9 1123 109

View Full Product Details