Assay Design



Gene Transcripts

Gene Symbol
asap2b
Entrez Gene ID
560581
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 2b
Gene Aliases
fe36f09, si:dkey-81b15.3, wu:fe36f09, zgc:158353
Chromosome Location
Chr.20: 29634611 - 29675457 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044933.2 NP_001038398.2 25 - 26 3038 107
GenBank mRNA BC129209.1 - 25 - 26 3038 107

View Full Product Details