Assay Design



Gene Transcripts

Gene Symbol
otud5a
Entrez Gene ID
555459
Gene Name
OTU deubiquitinase 5a
Gene Aliases
DUBA, otud5, si:bz30i22.1, si:rp71-30i22.1, zgc:154072
Chromosome Location
Chr.8: 8860927 - 8922631 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075110.1 NP_001068578.1 8 - 9 1739 70
GenBank mRNA BC124533.1 - 8 - 9 1739 70
BC155304.1 - 9 - 10 1656 70

View Full Product Details